Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis)
Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis)
Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis)
Thermo Scientific Chemicals

Niobium wire, 1.0mm (0.04in) dia, 99.8% (metals basis)

Form: Approx.6.7g/m | CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
25 m
Catalog number 010264.G1
also known as 010264-G1
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.50
Save 7.85 (15%)
Each
Quantity:
1 m
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.50
Save 7.85 (15%)
Each
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 1.0mm (0.04in) diameter.
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.8% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only