Niobium wire, 0.25mm (0.01in) dia, 99.99% (metals basis excluding Ta)
Niobium wire, 0.25mm (0.01in) dia, 99.99% (metals basis excluding Ta)
Niobium wire, 0.25mm (0.01in) dia, 99.99% (metals basis excluding Ta)
Thermo Scientific Chemicals

Niobium wire, 0.25mm (0.01in) dia, 99.99% (metals basis excluding Ta)

CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
500 cm
Catalog number 010265.BU
also known as 010265-BU
Price (USD)
59.70
Each
Quantity:
25 cm
Request bulk or custom format
Price (USD)
59.70
Each
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Trace MetalTotal metal impurities less than 0.01% by weight excluding Ta
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only