Niobium rod, 6.35mm (0.25in) dia, annealed, 99.8% (metals basis)
Niobium rod, 6.35mm (0.25in) dia, annealed, 99.8% (metals basis)
Niobium rod, 6.35mm (0.25in) dia, annealed, 99.8% (metals basis)
Thermo Scientific Chemicals

Niobium rod, 6.35mm (0.25in) dia, annealed, 99.8% (metals basis)

CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 cm
25 cm
100 cm
Catalog number 010273.BQ
also known as 010273-BQ
Price (USD)
54.65
Special offer
Online exclusive
Ends: 28-Dec-2026
64.70
Save 10.05
Each
Quantity:
5 cm
Request bulk or custom format
Price (USD)
54.65
Special offer
Online exclusive
Ends: 28-Dec-2026
64.70
Save 10.05
Each
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.8% (metals basis)
FormRod, 6.35mm (0.25in) diameter, annealed.
Appearance (Color)Silvery
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only