Palladium wire, 0.25mm (0.01in) dia, 99.9% (metals basis)
Palladium wire, 0.25mm (0.01in) dia, 99.9% (metals basis)
Palladium wire, 0.25mm (0.01in) dia, 99.9% (metals basis)
Thermo Scientific Chemicals

Palladium wire, 0.25mm (0.01in) dia, 99.9% (metals basis)

Form: Approx.0.6g/m | CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
Catalog number 010278.G1
also known as 010278-G1
Price (USD)
-
Quantity:
1 m
Request bulk or custom format
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver white
FormWire, 0.25mm (0.01in) diameter.
Assay from Supplier's CofA≥99.9% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only