Platinum wire, 0.127mm (0.005in) dia, For ISA Type R or S Standard Grade Thermocouple
Platinum wire, 0.127mm (0.005in) dia, For ISA Type R or S Standard Grade Thermocouple
Platinum wire, 0.127mm (0.005in) dia, For ISA Type R or S Standard Grade Thermocouple
Thermo Scientific Chemicals

Platinum wire, 0.127mm (0.005in) dia, For ISA Type R or S Standard Grade Thermocouple

For ISA Type R or S Standard Grade Thermocouple | CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
25 m
Catalog number 010289.G1
also known as 010289-G1
Price (USD)
-
Quantity:
1 m
Request bulk or custom format
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentFor ISA Type R or S Standard Grade Thermocouple
FormWire, 0.127mm (0.005in) diameter
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only