Rhenium wire, 0.25mm (0.01in) dia, 99.97% (metals basis)
Rhenium wire, 0.25mm (0.01in) dia, 99.97% (metals basis)
Rhenium wire, 0.25mm (0.01in) dia, 99.97% (metals basis)
Thermo Scientific Chemicals

Rhenium wire, 0.25mm (0.01in) dia, 99.97% (metals basis)

CAS: 7440-15-5 | Re | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
50 cm
250 cm
Catalog number 010311.CB
also known as 010311-CB
Price (USD)
873.65
Online Exclusive
970.00
Save 96.35 (10%)
Each
Quantity:
250 cm
Request bulk or custom format
Price (USD)
873.65
Online Exclusive
970.00
Save 96.35 (10%)
Each
Chemical Identifiers
CAS7440-15-5
IUPAC Namerhenium
Molecular FormulaRe
InChI KeyWUAPFZMCVAUBPE-UHFFFAOYSA-N
SMILES[Re]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.25mm (0.01in) diameter
Total Metal Impurities≤0.03%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only