Rhenium powder, -325 mesh, 99.99% (metals basis)
Rhenium powder, -325 mesh, 99.99% (metals basis)
Rhenium powder, -325 mesh, 99.99% (metals basis)
Thermo Scientific Chemicals

Rhenium powder, -325 mesh, 99.99% (metals basis)

CAS: 7440-15-5 | Re | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.2 g
2 g
10 g
50 g
Catalog number 010313.09
also known as 010313-09
Price (USD)
571.65
Special offer
Online exclusive
Ends: 30-Jun-2026
672.00
Save 100.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
571.65
Special offer
Online exclusive
Ends: 30-Jun-2026
672.00
Save 100.35 (15%)
Each
Chemical Identifiers
CAS7440-15-5
IUPAC Namerhenium
Molecular FormulaRe
InChI KeyWUAPFZMCVAUBPE-UHFFFAOYSA-N
SMILES[Re]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentGray
Total Metal Impurities≤ 0.01%
FormPowder
Particle Size-325 Mesh
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only