Rubidium, 99.75% (metals basis)
Rubidium, 99.75% (metals basis)
Rubidium, 99.75% (metals basis)
Thermo Scientific Chemicals

Rubidium, 99.75% (metals basis)

CAS: 7440-17-7 | Rb | 85.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Catalog number 010315.06
Price (USD)
296.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
296.00
Each
Chemical Identifiers
CAS7440-17-7
IUPAC Namerubidium
Molecular FormulaRb
InChI KeyIGLNJRXAVVLDKE-UHFFFAOYSA-N
SMILES[Rb]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver-white
Total Metal Impurities≤ 0.25%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What type of container is Rubidium 99.75% (metal basis) sold in?

Cat. No. 044214.xx Rubidium 99.75% (metal basis) is ampouled under Argon.

Cat. No. 010315.xx Rubidium 99.75% (metal basis) is vacuum sealed in a breakseal ampoule.