Tantalum rod, 6.4mm (0.25in) dia, 99.95% (metals basis)
Tantalum rod, 6.4mm (0.25in) dia, 99.95% (metals basis)
Tantalum rod, 6.4mm (0.25in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals

Tantalum rod, 6.4mm (0.25in) dia, 99.95% (metals basis)

CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 cm
25 cm
Catalog number 010346.BQ
also known as 010346-BQ
Price (USD)
189.65
Online Exclusive
211.00
Save 21.35 (10%)
Each
Quantity:
5 cm
Request bulk or custom format
Price (USD)
189.65
Online Exclusive
211.00
Save 21.35 (10%)
Each
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod, 6.4mm (0.25in) diameter
Total Metal Impurities≤0.05%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only