Tantalum wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis)
Tantalum wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis)
Tantalum wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis)
Thermo Scientific™

Tantalum wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis)

Form: approx. 3.26g/m; Temper: soft | CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
25 m
Catalog number 010349.G9
also known as 010349-G9
Price (USD)
587.65
Online Exclusive
653.00
Save 65.35 (10%)
Each
Quantity:
25 m
Request bulk or custom format
Price (USD)
587.65
Online Exclusive
653.00
Save 65.35 (10%)
Each
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.5 mm (0.02 in) dia.
Total Metal Impurities≤0.05%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only