Tantalum foil, 0.5mm (0.02in) thick, annealed, 99.95% (metals basis excluding Nb)
Tantalum foil, 0.5mm (0.02in) thick, annealed, 99.95% (metals basis excluding Nb)
Tantalum foil, 0.5mm (0.02in) thick, annealed, 99.95% (metals basis excluding Nb)
Thermo Scientific Chemicals

Tantalum foil, 0.5mm (0.02in) thick, annealed, 99.95% (metals basis excluding Nb)

Form: approx. 5.19g/25x25mm | CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
200 x 200 mm
Catalog number 010355.GW
also known as 010355-GW
Price (USD)
2,056.65
Online Exclusive
2,285.00
Save 228.35 (10%)
Each
Quantity:
200 x 200 mm
Request bulk or custom format
Price (USD)
2,056.65
Online Exclusive
2,285.00
Save 228.35 (10%)
Each
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.05%, excluding Nb
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only