Tantalum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
Tantalum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
Tantalum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Tantalum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)

Form: Approx. 10.4g/25 x 25mm | CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 010356.FI
also known as 010356-FI
Price (USD)
318.65
Special offer
Online exclusive
Ends: 28-Dec-2026
375.00
Save 56.35
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
318.65
Special offer
Online exclusive
Ends: 28-Dec-2026
375.00
Save 56.35
Each
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 1.0mm (0.04in) thick.
Assay from Supplier's CofA99.95% (metals basis)
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.95% (metals basis)
FormFoil, 1.0mm (0.04in) thick, hardened.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only