Tin powder, -325 mesh
Tin powder, -325 mesh
Tin powder, -325 mesh
Tin powder, -325 mesh
Thermo Scientific Chemicals

Tin powder, -325 mesh

Apparent density 3.2g/cm3 | CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 010379.36
also known as 010379-36
Price (USD)
123.65
Online Exclusive
137.00
Save 13.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
123.65
Online Exclusive
137.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size≥98.0% -325 mesh
DensityApparent Density: 2.9-3.5 g/cc
FormPowder, -325 mesh
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only