Titanium wire, 1.0mm (0.04in) dia, annealed, 99.99% (metals basis)
Titanium wire, 1.0mm (0.04in) dia, annealed, 99.99% (metals basis)
Titanium wire, 1.0mm (0.04in) dia, annealed, 99.99% (metals basis)
Thermo Scientific Chemicals

Titanium wire, 1.0mm (0.04in) dia, annealed, 99.99% (metals basis)

Form: Approx. 3.54g/m | CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
500 cm
Catalog number 010388.CC
also known as 010388-CC
Price (USD)
573.65
Special offer
Online exclusive
Ends: 30-Jun-2026
675.00
Save 101.35 (15%)
Each
Quantity:
500 cm
Request bulk or custom format
Price (USD)
573.65
Special offer
Online exclusive
Ends: 30-Jun-2026
675.00
Save 101.35 (15%)
Each
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 1.0mm (0.04in) dia, annealed
Total Metal Impurities≤0.01%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only