Titanium rod, 6.4mm (0.25in) dia, 99.7% (metals basis)
Titanium rod, 6.4mm (0.25in) dia, 99.7% (metals basis)
Titanium rod, 6.4mm (0.25in) dia, 99.7% (metals basis)
Thermo Scientific Chemicals

Titanium rod, 6.4mm (0.25in) dia, 99.7% (metals basis)

Form: Approx. 1.45g/cm | CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
5 x 100 cm
Catalog number 010393.CQ
also known as 010393-CQ
Price (USD)
371.65
Online Exclusive
413.00
Save 41.35 (10%)
Each
Quantity:
5 x 100 cm
Request bulk or custom format
Price (USD)
371.65
Online Exclusive
413.00
Save 41.35 (10%)
Each
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.3% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only