Titanium foil, 0.89mm (0.035in) thick, annealed, 99.7% (metals basis)
Titanium foil, 0.89mm (0.035in) thick, annealed, 99.7% (metals basis)
Titanium foil, 0.89mm (0.035in) thick, annealed, 99.7% (metals basis)
Titanium foil, 0.89mm (0.035in) thick, annealed, 99.7% (metals basis)
Thermo Scientific Chemicals

Titanium foil, 0.89mm (0.035in) thick, annealed, 99.7% (metals basis)

Form: Approx. 40g/100 x 100mm | CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
200 x 200 mm
200 x 1000 mm
Catalog number 010399.GH
also known as 010399-GH
Price (USD)
46.65
Online exclusive
52.20
Save 5.55
Each
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)
46.65
Online exclusive
52.20
Save 5.55
Each
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.89mm (0.035in) thick, annealed
Total Metal Impurities≤0.3%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only