Tungsten wire, 0.025mm (0.001in) dia, 99.95% (metals basis)
Tungsten wire, 0.025mm (0.001in) dia, 99.95% (metals basis)
Tungsten wire, 0.025mm (0.001in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals

Tungsten wire, 0.025mm (0.001in) dia, 99.95% (metals basis)

Form: Approx. 0.009g/m | CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Catalog NumberQuantity
010405.H4
also known as 010405-H4
100 m
Catalog number 010405.H4
also known as 010405-H4
Price (USD)
194.65
Special offer
Online exclusive
Ends: 30-Jun-2026
229.00
Save 34.35 (15%)
Each
Quantity:
100 m
Request bulk or custom format
Price (USD)
194.65
Special offer
Online exclusive
Ends: 30-Jun-2026
229.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS7440-33-7
IUPAC Nametungsten
Molecular FormulaW
InChI KeyWFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES[W]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
Assay from Supplier's CofA≥99.95% (metals basis)
FormWire, 0.025mm (0.001in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only