Tungsten rod, 6.3mm (0.25in) dia, annealed, 99.95% (metals basis)
Tungsten rod, 6.3mm (0.25in) dia, annealed, 99.95% (metals basis)
Tungsten rod, 6.3mm (0.25in) dia, annealed, 99.95% (metals basis)
Thermo Scientific Chemicals

Tungsten rod, 6.3mm (0.25in) dia, annealed, 99.95% (metals basis)

CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
50 cm
Catalog number 010406.BS
also known as 010406-BS
Price (USD)
114.65
Online Exclusive
127.00
Save 12.35 (10%)
Each
Quantity:
10 cm
Request bulk or custom format
Price (USD)
114.65
Online Exclusive
127.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS7440-33-7
IUPAC Nametungsten
Molecular FormulaW
InChI KeyWFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES[W]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
FormRod, 6.3mm (0.25in) diameter, annealed.
Assay from Supplier's CofA≥99.95% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only