Tungsten foil, 0.5mm (0.02in) thick, 99.95% (metals basis)
Tungsten foil, 0.5mm (0.02in) thick, 99.95% (metals basis)
Tungsten foil, 0.5mm (0.02in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Tungsten foil, 0.5mm (0.02in) thick, 99.95% (metals basis)

Form: Approx. 12g/25 x 50mm | CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 50 mm
50 x 100 mm
50 x 500 mm
200 x 200 mm
300 x 300 mm
Catalog number 010414.FG
also known as 010414-FG
Price (USD)
117.65
Online Exclusive
131.00
Save 13.35 (10%)
Each
Quantity:
25 x 50 mm
Request bulk or custom format
Price (USD)
117.65
Online Exclusive
131.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS7440-33-7
IUPAC Nametungsten
Molecular FormulaW
InChI KeyWFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES[W]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.5mm (0.02in) thick
Total Metal Impurities≤0.05%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only