Di-^m-chlorobis(norbornadiene)dirhodium(I), Rh 44% min
Di-^m-chlorobis(norbornadiene)dirhodium(I), Rh 44% min
Di-^m-chlorobis(norbornadiene)dirhodium(I), Rh 44% min
Thermo Scientific Chemicals

Di-^m-chlorobis(norbornadiene)dirhodium(I), Rh 44% min

CAS: 12257-42-0 | C14H16Cl2Rh2 | 460.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
1 g
Catalog number 010467.02
also known as 010467-02
Price (USD)
-
Quantity:
0.5 g
Request bulk or custom format
Chemical Identifiers
CAS12257-42-0
IUPAC Namebis(λ¹-rhodium(1+)) bis(bicyclo[2.2.1]hepta-2,5-diene) dichloride
Molecular FormulaC14H16Cl2Rh2
InChI KeyRXDWVIOULPVOEO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Rh+].[Rh+].C1C2C=CC1C=C2.C1C2C=CC1C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)35.50-37.50% Carbon
Assay (unspecified)3.00-4.00% Hydrogen
Assay (unspecified)Rh 43.6-45.6% min
Di-μ-chlorobis(norbornadiene)dirhodium(I) is used as catalysts in olefin hydroformylation, olefin hydrogenation and olefin isomerization

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only