Chlorotris(triphenylphosphine)rhodium(I), 97%
Chlorotris(triphenylphosphine)rhodium(I), 97%
Chlorotris(triphenylphosphine)rhodium(I), 97%
Thermo Scientific Chemicals

Chlorotris(triphenylphosphine)rhodium(I), 97%

CAS: 14694-95-2 | C54H45ClP3Rh | 925.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 010468.03
also known as 010468-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS14694-95-2
IUPAC Nameλ¹-rhodium(1+) tris(triphenylphosphane) chloride
Molecular FormulaC54H45ClP3Rh
InChI KeyIXAYKDDZKIZSPV-UHFFFAOYSA-M
SMILES[Cl-].[Rh+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.1% min
Chlorotris(triphenylphosphine)rhodium(I) is used as a homogeneous hydrogenation catalyst. It is used in the selective hydrogenation of alkenes and alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only