cis-Diamminedichloroplatinum(II), Pt 64.5% min
cis-Diamminedichloroplatinum(II), Pt 64.5% min
cis-Diamminedichloroplatinum(II), Pt 64.5% min
Thermo Scientific Chemicals

cis-Diamminedichloroplatinum(II), Pt 64.5% min

Potent Anticancer Agent That Blocks DNA Synthesis. | CAS: 15663-27-1 | Cl2H6N2Pt | 300.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number 010471.01
also known as 010471-01
Price (USD)
-
Quantity:
0.25 g
Request bulk or custom format
Chemical Identifiers
CAS15663-27-1
IUPAC Namedichloroplatinumbis(ylium) diamine
Molecular FormulaCl2H6N2Pt
InChI KeyBSJGASKRWFKGMV-UHFFFAOYSA-L
SMILESN.N.Cl[Pt++]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pt 64.5% min
Being a potent anticancer agent that blocks DNA synthesis, the pharmaceutical compositions are used in chemotherapy. First member of platinum-containing anti-cancer drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only