trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min
trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min
trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min
Thermo Scientific Chemicals

trans-Dichlorobis(triphenylphosphine)palladium(II), Pd 14.0% min

Hydrogenation, hydrosilation, carbonylation, oxidation, and C-C bond formation | CAS: 13965-03-2 | C36H30Cl2P2Pd | 701.90 g/mol
Have Questions?
Catalog NumberQuantity
010491.03
also known as 010491-03
1 g
Catalog number 010491.03
also known as 010491-03
Price (USD)
47.65
Online exclusive
53.00
Save 5.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
47.65
Online exclusive
53.00
Save 5.35
Each
Chemical Identifiers
CAS13965-03-2
IUPAC Namepalladium(2+) bis(triphenylphosphane) dichloride
Molecular FormulaC36H30Cl2P2Pd
InChI KeyYNHIGQDRGKUECZ-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pd 14.0% min
As a catalyst of choice in hydrogenation, hydrosilation, carbonylation, oxidation, elimination, reduction, allyl aceate isomerization, dimerization of 1,3-dienes, and C-C bond formation such as Negishi coupling, Suzuki coupling, Kumada coupling, Sonogashira coupling, Heck coupling and Sonogashira-Hagihara coupling reactions. Used in the preparation of tetrakis(triphenylphosphine)palladium, condensed heterocycles such as furoquinolines, diphenylacetylenes, and several other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only