Tricarbonyldichlororuthenium(II) dimer
Tricarbonyldichlororuthenium(II) dimer
Tricarbonyldichlororuthenium(II) dimer
Thermo Scientific Chemicals

Tricarbonyldichlororuthenium(II) dimer

CAS: 22594-69-0 | C6Cl4O6Ru2 | 512.00 g/mol
Have Questions?
Catalog NumberQuantity
010503.03
also known as 010503-03
1 g
Catalog number 010503.03
also known as 010503-03
Price (USD)
292.65
Online Exclusive
325.00
Save 32.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
292.65
Online Exclusive
325.00
Save 32.35 (10%)
Each
Chemical Identifiers
CAS22594-69-0
IUPAC Namebis(λ²-ruthenium(2+)) hexakis(methanidylidyneoxidanium) tetrachloride
Molecular FormulaC6Cl4O6Ru2
InChI KeyJYHHJVKGDCZCCL-UHFFFAOYSA-J
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Ru++].[Ru++].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98% min
Tricarbonyldichlororuthenium(II) dimer is used to enhance the coagulation and attenuation of vulnerability to fibrinolysis, modify thrombus growth or disintegration, enhances fibrinogen as a substrate for thrombin, and regulation of ion transport by gasotransmitters. Also used for various pharmacological studies. It is used as a CO donor for reactive oxygen species mediated bacterial killing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only