Thermo Scientific Chemicals

Iridium(III) 2,4-pentanedionate, Ir 37.5% min, Thermo Scientific Chemicals

Catalog number: 010514.03
1 g, Each
Thermo Scientific Chemicals

Iridium(III) 2,4-pentanedionate, Ir 37.5% min, Thermo Scientific Chemicals

Catalog number: 010514.03
1 g, Each
Quantity
Catalog number: 010514.03
also known as 010514-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15635-87-7
IUPAC Name
iridium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular Formula
C15H21IrO6
InChI Key
HLYTZTFNIRBLNA-LNTINUHCSA-K
SMILES
[Ir+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Assay (unspecified)
Ir 37.5% min

Description

It is used as a photoreducing agent, a precursor for preparing Ir-C thin film and a catalyst for hydrogenation of sulfur containing compounds. Used in various catalysis reactions in organic synthesis, including carbon nanostructures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text