What storage conditions should I use for Lithium chloride?
Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.
Search
View additional product information for Lithium chloride, anhydrous, 99% - FAQs (010515.A1, 010515.30, 010515.A7)
1 product FAQs found