Potassium tetracyanopalladate(II) hydrate
Potassium tetracyanopalladate(II) hydrate
Potassium tetracyanopalladate(II) hydrate
Thermo Scientific Chemicals

Potassium tetracyanopalladate(II) hydrate

CAS: 14516-46-2 | C4K2N4Pd | 288.69 g/mol
Have Questions?
Catalog NumberQuantity
010535.03
also known as 010535-03
1 g
Catalog number 010535.03
also known as 010535-03
Price (USD)
197.65
Online Exclusive
219.00
Save 21.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
197.65
Online Exclusive
219.00
Save 21.35 (10%)
Each
Chemical Identifiers
CAS14516-46-2
IUPAC Namedipotassium palladiumtetracarbonitrile
Molecular FormulaC4K2N4Pd
InChI KeyNKYOTIKKMTWAOI-UHFFFAOYSA-N
SMILES[K+].[K+].N#C[Pd](C#N)(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pd 30% min

Potassium tetracyanopalladate(II) hydrate is used as a primary and secondary intermediate in Chemical & pharmaceutical research. it is also used as catalysts, as precursors of metallic palladium, in preparative chemical production, in photography, in electroplating and medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only