Thermo Scientific Chemicals

Rubidium carbonate, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 010562.09
10 g, Each
Thermo Scientific Chemicals

Rubidium carbonate, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 010562.09
10 g, Each
Quantity
Catalog number: 010562.09
also known as 010562-09
Price (USD)
Price: 66.30
Online price: 56.65
Your price:
Quantity
-

Chemical Identifiers

CAS
584-09-8
IUPAC Name
dirubidium(1+) carbonate
Molecular Formula
CO3Rb2
InChI Key
WPFGFHJALYCVMO-UHFFFAOYSA-L
SMILES
[Rb+].[Rb+].[O-]C([O-])=O
Appearance (Color)
White
Form
Crystals or crystalline powder or powder
Total Metal Impurities
≤0.20% (≤2000 ppm)

Description

Rubidium carbonate is used in glass-making by enhancing stability and durability as well as reducing its conductivity. It is also used as a part of a catalyst for preparing short-chain alcohols from feed gas. Rubidium carbonate glass has been extensively tested for use in anti-collision devices for motor vehicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text