Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis)
Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis)
Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis)
Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis)
Thermo Scientific Chemicals

Rubidium hydroxide, 50% w/w aq. soln., 99.6+% (metals basis)

CAS: 1310-82-3 | HORb | 102.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 010565.NC
also known as 010565-NC
Price (USD)
141.65
Online Exclusive
157.00
Save 15.35 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
141.65
Online Exclusive
157.00
Save 15.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1310-82-3
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.4% max.
Rubidium Hydroxide is used to give fireworks a violet color. It is a notable ntermediate in the synthesis of other rubidium compounds. It is used in photography and storage batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only