Nickel foil, 0.127mm (0.005in) thick, annealed, 99+% (metals basis)
Nickel foil, 0.127mm (0.005in) thick, annealed, 99+% (metals basis)
Nickel foil, 0.127mm (0.005in) thick, annealed, 99+% (metals basis)
Nickel foil, 0.127mm (0.005in) thick, annealed, 99+% (metals basis)
Thermo Scientific Chemicals

Nickel foil, 0.127mm (0.005in) thick, annealed, 99+% (metals basis)

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
20 x 30 cm
30 x 100 cm
30 x 500 cm
Catalog number 010595.CH
also known as 010595-CH
Price (USD)
118.00
Each
Quantity:
20 x 30 cm
Request bulk or custom format
Price (USD)
118.00
Each
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only