Aluminum chloride hexahydrate, Puratronic™, 99.9995% (metals basis)
Aluminum chloride hexahydrate, Puratronic™, 99.9995% (metals basis)
Aluminum chloride hexahydrate, Puratronic™, 99.9995% (metals basis)
Thermo Scientific Chemicals

Aluminum chloride hexahydrate, Puratronic™, 99.9995% (metals basis)

CAS: 7784-13-6 | AlCl3H12O6 | 241.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
Catalog number 010622.06
also known as 010622-06
Price (USD)
62.65
Online exclusive
69.70
Save 7.05
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
62.65
Online exclusive
69.70
Save 7.05
Each
Chemical Identifiers
CAS7784-13-6
IUPAC Namealuminium(3+) hexahydrate trichloride
Molecular FormulaAlCl3H12O6
InChI KeyJGDITNMASUZKPW-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder
Total Metal Impurities≤0.0005% (5 ppm)
Appearance (Color)White
Assay (special)?2.5 % of Theoretical value (Al)
Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only