Thermo Scientific Chemicals

Antimony(V) oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010643.22
100 g, Each
Thermo Scientific Chemicals

Antimony(V) oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010643.22
100 g, Each
Quantity
Catalog number: 010643.22
also known as 010643-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
1314-60-9
IUPAC Name
[(dioxostibanyl)oxy]stibanedione
Molecular Formula
O5Sb2
InChI Key
LJCFOYOSGPHIOO-UHFFFAOYSA-N
SMILES
O=[Sb](=O)O[Sb](=O)=O
Appearance (Color)
Light Yellow
Total Metal Impurities
≤0.002% (20 ppm)
Assay (special)
?2.5 % of Theoretical value (Sb)
Form
Powder

Description

It is used as flame retardants in acrylonitrile-butadiene-styrene (ABS) polymer, plastics, rubbers, paper and textiles. It finds use as a flocculant in the production of titanium dioxide. It also finds use in ion-exchange resin. It is also employed as catalyst in certain polymerization and oxidation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text