Bismuth(III) oxide, Puratronic™, 99.9995% (metals basis)
Bismuth(III) oxide, Puratronic™, 99.9995% (metals basis)
Bismuth(III) oxide, Puratronic™, 99.9995% (metals basis)
Thermo Scientific Chemicals

Bismuth(III) oxide, Puratronic™, 99.9995% (metals basis)

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 010658.18
also known as 010658-18
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Chemical Identifiers
CAS1304-76-3
IUPAC Namedibismuth(3+) trioxidandiide
Molecular FormulaBi2O3
InChI KeyTYIXMATWDRGMPF-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Bi+3].[Bi+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0005% max.
It is used as a precursor to the preparation of other compounds of bismuth, chemical reagents. It has specialized uses in optical glass, flame-retardant paper, in glaze formulations as a substitute for lead oxides. It is used as solid electrolyte because of its high thermal expansion property. It is used in radio frequency shielding, compound for catheters and other medical tubing products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only