Boric acid, Puratronic™, 99.9995% (metals basis)
Boric acid, Puratronic™, 99.9995% (metals basis)
Boric acid, Puratronic™, 99.9995% (metals basis)
Thermo Scientific Chemicals

Boric acid, Puratronic™, 99.9995% (metals basis)

CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010659.14
also known as 010659-14
Price (USD)
59.60
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
59.60
Each
Chemical Identifiers
CAS10043-35-3
IUPAC Nameboric acid
Molecular FormulaBH3O3
InChI KeyKGBXLFKZBHKPEV-UHFFFAOYSA-N
SMILESOB(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.0005% (5 ppm)
Assay (special)?2.5 % of Theoretical value (B)
FormCrystals or powder
Appearance (Color)White
It is used as a primary or adjunct buffer in swimming pools, flame retardant, neutron absorber in nuclear power plants, colorant to make flame green, preservative in urine samples, welding flux in combination with borax, or precursor to other chemical compounds. It is used to prevent/remove existing wet and dry rot in timbers, treat wood against fungal/insect attack, prevent athlete’s foot, lubricate carom boards, and reduce surface oxidation on metals at high temperatures. It can be used to produce lubricant for ceramic or metal surfaces, textile fiberglass, induction furnace linings and ceramics, Silly Putty, and glass LCD flat panel displays. It can prevent the high temperature reaction of aluminum with nitrates in pyrotechniques, and neutralize active hydrofluoric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only