Cadmium carbonate, Puratronic™, 99.998% (metals basis)
Cadmium carbonate, Puratronic™, 99.998% (metals basis)
Cadmium carbonate, Puratronic™, 99.998% (metals basis)
Thermo Scientific Chemicals

Cadmium carbonate, Puratronic™, 99.998% (metals basis)

CAS: 513-78-0 | CCdO3 | 172.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010660.09
also known as 010660-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS513-78-0
IUPAC Namecadmium(2+) carbonate
Molecular FormulaCCdO3
InChI KeyGKDXQAKPHKQZSC-UHFFFAOYSA-L
SMILES[Cd++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Cadmium carbonate is the raw material for the production of pigments for ceramic and frits and for production of cadmium salt. Used as reagents. It is used for intermediates of manufacturing polyester, glass pigment flux, organic catalysts, plastic plasticizer, stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only