Cadmium oxide, Puratronic™, 99.998% (metals basis)
Cadmium oxide, Puratronic™, 99.998% (metals basis)
Cadmium oxide, Puratronic™, 99.998% (metals basis)
Thermo Scientific Chemicals

Cadmium oxide, Puratronic™, 99.998% (metals basis)

CAS: 1306-19-0 | CdO | 128.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010664.14
also known as 010664-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1306-19-0
IUPAC Namecadmium(2+) oxidandiide
Molecular FormulaCdO
InChI KeyCFEAAQFZALKQPA-UHFFFAOYSA-N
SMILES[O--].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Cadmium oxide is used in ceramic glazes, cadmium electroplating baths, pigments, phosphors, electrodes for storage batteries, cadmium salts, and heterogeneous catalysis for dehydrogenation. It is a basic conducting material used to prepare transparent conducting films, which finds use in phototransistors, liquid crystal displays, photodiodes, photovoltaic cells, and antireflection coatings. Electrodeposited cadmium oxide films have excellent electrocatalytic activity towards many organic compounds, ozone and oxygen evolution. CdO is selective in phenol photodegradation. Glassy carbon electrode coated with cadmium oxide film has been used to develop a sensitive and rapid assay method for penicillamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only