Platinum powder, -22 mesh, Premion™, 99.99% (metals basis)
Platinum powder, -22 mesh, Premion™, 99.99% (metals basis)
Platinum powder, -22 mesh, Premion™, 99.99% (metals basis)
Thermo Scientific Chemicals

Platinum powder, -22 mesh, Premion™, 99.99% (metals basis)

CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number 010672.03
also known as 010672-03
Price (USD)
336.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
336.00
Each
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
FormPowder
Total Metal Impurities≤0.01%
Particle Size-22 mesh
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only