Cadmium sulfate hydrate, Puratronic™, 99.996% (metals basis)
Cadmium sulfate hydrate, Puratronic™, 99.996% (metals basis)
Cadmium sulfate hydrate, Puratronic™, 99.996% (metals basis)
Thermo Scientific Chemicals

Cadmium sulfate hydrate, Puratronic™, 99.996% (metals basis)

CAS: 15244-35-6 | CdO4S | 208.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010675.09
also known as 010675-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS15244-35-6
IUPAC Namecadmium(2+) sulfate
Molecular FormulaCdO4S
InChI KeyQCUOBSQYDGUHHT-UHFFFAOYSA-L
SMILES[Cd++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.004% max.
It is used as an electrolyte in a Weston standard cell, a pigment in fluorescent screens. It is used in the manufacture of cadmium salts of fatty acids, pigments, in electroplating of cadmium in electronic circuits. It act as an intermediate, as an accelerator, in the electrode position of nickel and copper, for stabilizing plastics, in cement formation. It has the ability to absorb hydrochloric acid from waste gases from chemical plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only