Calcium carbonate, Puratronic™, 99.99% (metals basis)
Calcium carbonate, Puratronic™, 99.99% (metals basis)
Calcium carbonate, Puratronic™, 99.99% (metals basis)
Calcium carbonate, Puratronic™, 99.99% (metals basis)
Thermo Scientific Chemicals

Calcium carbonate, Puratronic™, 99.99% (metals basis)

CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 010679.36
also known as 010679-36
Price (USD)
1,918.65
Online Exclusive
2,132.00
Save 213.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
1,918.65
Online Exclusive
2,132.00
Save 213.35 (10%)
Each
Chemical Identifiers
CAS471-34-1
IUPAC Namecalcium carbonate
Molecular FormulaCCaO3
InChI KeyVTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES[Ca++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.01% (100 ppm)
Assay (special)?2.5 % of Theoretical value (Ca)
FormPowder
Appearance (Color)White
Calcium carbonate is used in medicine and agriculture. It is also used in flue gas desulfurization in fossil fuel power stations, production of toothpaste, ceramics/glazing applications, purification of iron from iron ore, and to clean tarnish on silver. It forms the major component of ceramic tile adhesives. It is used in the refining of sugar, as filler in thermosetting resins, as an abrasive, as an extender in paints, and as a formation-bridging filtercake-sealing agent in oil industry. It is used in swimming pools for controlling the pH, to improve the rigidity of polypropylene compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only