Calcium fluoride, Puratronic™, 99.985% (metals basis)
Calcium fluoride, Puratronic™, 99.985% (metals basis)
Calcium fluoride, Puratronic™, 99.985% (metals basis)
Thermo Scientific Chemicals

Calcium fluoride, Puratronic™, 99.985% (metals basis)

CAS: 7789-75-5 | CaF2 | 78.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 010681.18
also known as 010681-18
Price (USD)
545.65
Special offer
Online exclusive
Ends: 30-Jun-2026
642.00
Save 96.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
545.65
Special offer
Online exclusive
Ends: 30-Jun-2026
642.00
Save 96.35 (15%)
Each
Chemical Identifiers
CAS7789-75-5
IUPAC Namecalcium difluoride
Molecular FormulaCaF2
InChI KeyWUKWITHWXAAZEY-UHFFFAOYSA-L
SMILES[F-].[F-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.015% max.
Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only