Chromium(III) oxide, Puratronic™, 99.97% (metals basis)
Chromium(III) oxide, Puratronic™, 99.97% (metals basis)
Chromium(III) oxide, Puratronic™, 99.97% (metals basis)
Thermo Scientific Chemicals

Chromium(III) oxide, Puratronic™, 99.97% (metals basis)

CAS: 1308-38-9 | Cr2O3 | 151.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 010688.06
also known as 010688-06
Price (USD)
61.30
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
61.30
Each
Chemical Identifiers
CAS1308-38-9
IUPAC Namedichromium(3+) trioxidandiide
Molecular FormulaCr2O3
InChI KeyUOUJSJZBMCDAEU-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Cr+3].[Cr+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.03% max.
Also known as green compound for buffing and polishing, it can be used as abrasives. Apart from being used as a precursor to the magnetic pigment viz., chromium dioxide, it is used in inks, glasses, and pigments (viridian). They are employed as plating agents and surface treating agents. It is a catalyst in the Phillips process of manufacture of high density polythene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only