Cobalt(II) chloride hexahydrate, Puratronic™, 99.998% (metals basis)
Cobalt(II) chloride hexahydrate, Puratronic™, 99.998% (metals basis)
Cobalt(II) chloride hexahydrate, Puratronic™, 99.998% (metals basis)
Thermo Scientific Chemicals

Cobalt(II) chloride hexahydrate, Puratronic™, 99.998% (metals basis)

CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010692.09
also known as 010692-09
Price (USD)
106.65
Special offer
Online exclusive
Ends: 28-Dec-2026
125.00
Save 18.35
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
106.65
Special offer
Online exclusive
Ends: 28-Dec-2026
125.00
Save 18.35
Each
Chemical Identifiers
CAS7791-13-1
IUPAC Nameλ²-cobalt(2+) hexahydrate dichloride
Molecular FormulaCl2CoH12O6
InChI KeyGFHNAMRJFCEERV-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Cobalt(II) chloride hexahydrate has been used to study apoptotic effects in HepG2 cells. Also reported as a stabilizer for reconstitution of the radiopharmaceutical technetium-99m exametazime. It has been used in studies to examine heat shock protein expression of inflamed skin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only