Cobalt(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis)
Cobalt(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis)
Cobalt(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Cobalt(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis)

CAS: 10026-24-1 | CoH14O11S | 281.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010696.14
also known as 010696-14
Price (USD)
197.65
Special offer
Online exclusive
Ends: 28-Dec-2026
232.00
Save 34.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
197.65
Special offer
Online exclusive
Ends: 28-Dec-2026
232.00
Save 34.35
Each
Chemical Identifiers
CAS10026-24-1
IUPAC Nameλ²-cobalt(2+) heptahydrate sulfate
Molecular FormulaCoH14O11S
InChI KeyMEYVLGVRTYSQHI-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Cobalt(II) sulfate heptahydrate is used in the preparation of pigments and other cobalt salts. It is used in storage batteries and electroplating baths, sympathetic inks, and as an additive to soils and animal feeds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only