Copper(II) sulfate hydrate, Puratronic™, 99.999% (metals basis)
Copper(II) sulfate hydrate, Puratronic™, 99.999% (metals basis)
Copper(II) sulfate hydrate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Copper(II) sulfate hydrate, Puratronic™, 99.999% (metals basis)

CAS: 23254-43-5 | CuO4S | 159.60 (anhydrous basis) g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010701.14
also known as 010701-14
Price (USD)
136.65
Special offer
Online exclusive
Ends: 28-Dec-2026
161.00
Save 24.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
Ends: 28-Dec-2026
161.00
Save 24.35
Each
Chemical Identifiers
CAS23254-43-5
IUPAC Namecopper(2+) sulfate
Molecular FormulaCuO4S
InChI KeyARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES[Cu++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
It is used as a catalyst in certain organic transformations like synthesis of ynamides. It is used in Fehling's solution and Benedict's solution for testing reducing sugars, in the Biuret reagent for testing proteins, and in combination with potassium permanganate for testing blood for anemia. It is used in the preparation of Cheshunt compound for horticulture. It finds use as a coloring ingredient in artworks, as an oxidant in combination with potassium permanganate for oxidizing primary alcohols, and as a mordant in dyeing. It is used to etch copper or zinc plates for intaglio print making.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only