Indium(I) chloride, Puratronic™, 99.999% (metals basis)
Indium(I) chloride, Puratronic™, 99.999% (metals basis)
Indium(I) chloride, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Indium(I) chloride, Puratronic™, 99.999% (metals basis)

CAS: 13465-10-6 | ClIn | 150.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 010707.06
also known as 010707-06
Price (USD)
311.65
Online Exclusive
346.00
Save 34.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
311.65
Online Exclusive
346.00
Save 34.35 (10%)
Each
Chemical Identifiers
CAS13465-10-6
IUPAC Nameλ¹-indiganyliumylidene chloride
Molecular FormulaClIn
InChI KeyAPHGZSBLRQFRCA-UHFFFAOYSA-M
SMILES[Cl-].[In+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Indium(I) chloride is used to prepare cage, dinuclear complexes (In-cyclodiphophazanes) which possess interesting metal-metal interactions, Semiconductor doping, phosphors-compound semiconductors, ITO thin-film, Basic raw material for compounding organic indium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only