Iron(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)
Iron(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)
Iron(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Iron(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)

CAS: 7782-61-8 | FeH18N3O18 | 403.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 010715.14
also known as 010715-14
Price (USD)
486.65
Online Exclusive
540.00
Save 53.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
486.65
Online Exclusive
540.00
Save 53.35 (10%)
Each
Chemical Identifiers
CAS7782-61-8
IUPAC Nameiron(3+) nonahydrate trinitrate
Molecular FormulaFeH18N3O18
InChI KeySZQUEWJRBJDHSM-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
It is used as catalyst for the synthesis of sodium amide from sodium and ammonia, and for etching of silver and silver alloys. Ferric nitrate is used in the synthesis of organically templated iron phosphates, and acts as an efficient catalyst in enamination of beta dicarbonyl compounds to beta enaminones. Ferric nitrate impregnated clays are useful as oxidizing agents. Clayfen, produced from ferric nitrate and montmorillonite, is used for the oxidation of thiols to disulfide, and alcohols to aldehydes. Ferric nitrate is useful for regio- and stereo-selective nitration of various aromatic, aliphatic, and heteroaromatic olefins. This reaction provides nitroolefins with excellent E-selectivity. With Keggin-type heteropoly acids, it selectively oxidizes various hydroxy groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only