Iron(II) oxalate dihydrate, Puratronic™, 99.999% (metals basis)
Iron(II) oxalate dihydrate, Puratronic™, 99.999% (metals basis)
Iron(II) oxalate dihydrate, Puratronic™, 99.999% (metals basis)
Iron(II) oxalate dihydrate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Iron(II) oxalate dihydrate, Puratronic™, 99.999% (metals basis)

CAS: 6047-25-2 | C2H4FeO6 | 179.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010718.22
also known as 010718-22
Price (USD)
137.65
Online Exclusive
153.00
Save 15.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
137.65
Online Exclusive
153.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS6047-25-2
IUPAC Nameλ²-iron(2+) dihydrate oxalate
Molecular FormulaC2H4FeO6
InChI KeyNPLZZSLZTJVZSX-UHFFFAOYSA-L
SMILESO.O.[Fe++].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Appearance (Color)yellow to orange
Formpowder
It is used as photographic developer for silver bromide-gelatin plates. It imparts greenish-brown tint to optical glass (sunglasses, windshields, railroad car windows), for decorative glassware, pigment for plastics, paints and lacquers. It acts as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only