Lithium chloride hydrate, Puratronic™, 99.996% (metals basis)
Lithium chloride hydrate, Puratronic™, 99.996% (metals basis)
Lithium chloride hydrate, Puratronic™, 99.996% (metals basis)
Thermo Scientific Chemicals

Lithium chloride hydrate, Puratronic™, 99.996% (metals basis)

CAS: 85144-11-2 | ClLi | 42.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010735.22
also known as 010735-22
Price (USD)
629.65
Online exclusive
699.00
Save 69.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
629.65
Online exclusive
699.00
Save 69.35
Each
Chemical Identifiers
CAS85144-11-2
IUPAC Namelithium(1+) chloride
Molecular FormulaClLi
InChI KeyKWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES[Li+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.004% max.
Lithium chloride is widely employed in lithium metal production. It is used as a brazing flux for aluminum in automobile parts. Its use can also be seen as a desiccant for drying air streams. In biochemical applications, lithium chloride is used to precipitate RNA from cellular extracts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What storage conditions should I use for Lithium chloride?

Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.