Lithium tetraborate, Puratronic™, 99.998% (metals basis)
Lithium tetraborate, Puratronic™, 99.998% (metals basis)
Lithium tetraborate, Puratronic™, 99.998% (metals basis)
Lithium tetraborate, Puratronic™, 99.998% (metals basis)
Thermo Scientific Chemicals

Lithium tetraborate, Puratronic™, 99.998% (metals basis)

CAS: 12007-60-2 | B4Li2O7 | 169.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Catalog number 010744.14
also known as 010744-14
Price (USD)
59.65
Online exclusive
66.60
Save 6.95
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
59.65
Online exclusive
66.60
Save 6.95
Each
Chemical Identifiers
CAS12007-60-2
IUPAC Namedilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Molecular FormulaB4Li2O7
InChI KeyRDZCISUGRQUANA-UHFFFAOYSA-N
SMILES[Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on ignition<2.0%
Total Metal Impurities≤0.002%
Appearance (Color)White
CommentDensity ca 0.55g/cm³
FormPowder
Used in the laboratory as LB (Lithium borate) buffer for gel electrophoresis of DNA and RNA. Employed in the borax fusion method to modify mineral powder samples for analysis by WDXRF (Wavelength Dispersive X-Ray Fluorescence) Spectroscopy. Acts as an ingredient in the manufacture of glasses and ceramics. Useful fod dissolving basic oxides such as CaO and MgO. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only