Magnesium acetate tetrahydrate, Puratronic™, 99.997% (metals basis)
Magnesium acetate tetrahydrate, Puratronic™, 99.997% (metals basis)
Magnesium acetate tetrahydrate, Puratronic™, 99.997% (metals basis)
Thermo Scientific Chemicals

Magnesium acetate tetrahydrate, Puratronic™, 99.997% (metals basis)

CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010795.14
also known as 010795-14
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
134.00
Save 20.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
113.65
Special offer
Online exclusive
Ends: 30-Jun-2026
134.00
Save 20.35 (15%)
Each
Chemical Identifiers
CAS16674-78-5
IUPAC Namemagnesium(2+) diacetate tetrahydrate
Molecular FormulaC4H14MgO8
InChI KeyXKPKPGCRSHFTKM-UHFFFAOYSA-L
SMILESO.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.003% max.
Used as a source of magnesium in biological reactions, and as a source of acetate ion in chemical transformations. Acts as an emission control agent in coal combustion process to reduce acid rain. It is involved in glass formation process via the thermal decomposition of solids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only